P-QUINONE AS CARBON 100 MG/L IN H2O 1 * 50 mL
Catalog # CMPLRD033612L05A
Supplier: CUSTOM MADE CHEMICALS LAB
P-QUINONE AS CARBON 100 MG/L IN H2O 1 * 50 mL
Catalog # CMPLRD033612L05A
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
VWR®, Test strips, chloride, Dosatest
Supplier: VWR Chemicals
Sodium hydroxide 35% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
VWR®, OCT Embedding matrix for frozen sections
Supplier: VWR Chemicals
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hypochlorite (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Accessories
Supplier: Hach