Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]MIXTURE 3 COMPONENTS 1 * 5 mL
Catalog # CMPLRD0911795
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
[EN]MIXTURE 3 COMPONENTS 1 * 5 mL
Catalog # CMPLRD0911795
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Supplier: Merck
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
[EN]FERROUS AMMONIUM SULPHATE 0.1M 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB