ContactUsLinkComponent
Thermostatically regulated hotplates, eloxidised aluminum heating surface, 350x350 mm, 1150W, 230V, 30...110C (Not suitable for cooking pursoses) 1 * 1 items
Catalog # GESTHT11
Supplier:  GESTIGKEIT HARRY
Thermostatically regulated hotplates, eloxidised aluminum heating surface, 350x350 mm, 1150W, 230V, 30...110C (Not suitable for cooking pursoses) 1 * 1 items
Catalog # GESTHT11
Supplier:  GESTIGKEIT HARRY
Frequently Bought Together