N-HEXADEKAN-D34 100 G/ML IN CYCLOHEXAN 1 * 5 mL
Catalog # RIVO147000361D100C
Supplier: RIVOLAB
Frequently Bought Together
Methyl benzenesulphonate
Supplier: USP
Cerium Standard for ICP 1 * 100 mL
Supplier: Merck
Conductivity standard 1413 µS/cm (25°C; 0,01 mol/L; KCl) for conductivity measurement
Supplier: SI Analytics
Supelco® TraceCERT® Organic Reference Standard, Tebutam
Supplier: Merck
Supelco® PESTANAL® Organic Reference Standard, Omethoate
Supplier: Merck
Buffer PH 4,00 1 * 500 mL
Supplier: REAGECON
Fonofos certified 1 * 0,1 g
Supplier: EHRENSTORFER
Nitrate standards for ISE calibration, Orion™
Supplier: Thermo Orion
Pesticide reference standard, 1,3-Dichlorobenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Supplier: EHRENSTORFER
Valeric acid, Supelco®
Supplier: Merck
Reference standard for food analysis, phthalic acid, Dr Ehrenstorfer
Supplier: EHRENSTORFER
D-Glucuronic Acid, Reference Standard
Supplier: USP