OTNE(Tetramethyl acetyloktahydronaftale)100 µg/ml in cyclohexane 1 * 5 mL
Catalog # RIVO134500115100CY
Supplier: RIVOLAB
OTNE(Tetramethyl acetyloktahydronaftale)100 µg/ml in cyclohexane 1 * 5 mL
Catalog # RIVO134500115100CY
Supplier: RIVOLAB
Frequently Bought Together
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
Conductivity standards, Certipur® Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solution, pH 9 (at 20 °C), Borate free
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Buffers, concentrated solutions, Titrisol®, Supelco®
Supplier: Merck
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
pH buffer, single-use, SINGLET™
Supplier: Hach