Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Vogel Bonner Agar, 9 cm 18 ml. The plates are packed per 10 pcs Shelf life 3 months. 1 * 10 items
Catalog # V222390136
Supplier:
BALIS
Expand image
Vogel Bonner Agar, 9 cm 18 ml. The plates are packed per 10 pcs Shelf life 3 months. 1 * 10 items
Catalog # V222390136
Supplier:
BALIS
Frequently Bought Together
Real Time PCR Detetction Test Kits, SUPREME
Supplier: BIOPREMIER
BRAIN HEART INFUSION AGAR 1 * 500 g
Supplier: LIOFILCHEM
VWR® Dehydrated Culture Media, Powdered
Supplier: VWR Chemicals
VWR®, Fluids and culture media for sterility testing of pharmaceutical products
Supplier: VWR Chemicals
Dehydrated culture media
Supplier: LIOFILCHEM
VWR® Dip Slides
Supplier: VWR Chemicals