Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Wipes Kitchen HACCP, yellow, viscose/polyester, suitable for food 1 * 300 items
Catalog # ECOLCOT3860Q06050Y
Supplier:
ECOLINE NV SA
Expand image
Wipes Kitchen HACCP, yellow, viscose/polyester, suitable for food 1 * 300 items
Catalog # ECOLCOT3860Q06050Y
Supplier:
ECOLINE NV SA
Frequently Bought Together
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Conductivity cells
Supplier: WTW
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
VWR® pHenomenal® pH Electrodes
Supplier: VWR Collection
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
Supplier: VWR Collection
pH electrodes, SenTix®
Supplier: WTW
VWR®, Latex Gloves
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Bench Protectors
Supplier: VWR Collection
Facial tissues, KLEENEX®
Supplier: KIMBERLY CLARK