ContactUsLinkComponent
5 components: Ammonium (NH4+) 100mg/l Potassium (K+) 100mg/l Sodium (Na+) 100mg/l Lithium (Li+) 20mg/l Ethanolamine (for Inorganics) (C2H7NO) 100mg/l in Water ,, CRM, ISO 17034 and ISO 17025,, Conservation 12 months. (6 months after opening), 1 * 500 mL
Catalog # CMPLRM092682L5
5 components: Ammonium (NH4+) 100mg/l Potassium (K+) 100mg/l Sodium (Na+) 100mg/l Lithium (Li+) 20mg/l Ethanolamine (for Inorganics) (C2H7NO) 100mg/l in Water ,, CRM, ISO 17034 and ISO 17025,, Conservation 12 months. (6 months after opening), 1 * 500 mL
Catalog # CMPLRM092682L5