ContactUsLinkComponent
1 component: PBB 101 [CAS:67888-96-4] 35mg/l in Iso-octane CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 1 mL
Catalog # CMPLRD0916121
1 component: PBB 101 [CAS:67888-96-4] 35mg/l in Iso-octane CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 1 mL
Catalog # CMPLRD0916121
Frequently Bought Together