ContactUsLinkComponent
26 components: 100mg/l each of Al Sb As Ba Be B Cd Ca Cr Co Cu Fe Pb Mg Mn Mo Ni K Se Si Ag Na Tl Ti V Zn in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Stor 1 * 100 mL
Catalog # CMPLRM092408L1
26 components: 100mg/l each of Al Sb As Ba Be B Cd Ca Cr Co Cu Fe Pb Mg Mn Mo Ni K Se Si Ag Na Tl Ti V Zn in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Stor 1 * 100 mL
Catalog # CMPLRM092408L1
Frequently Bought Together