CO 0.75MG/L IN HNO3 2% 1 * 100 mL
Catalog # CMPLRM091054L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Test kits, surfactants, Spectroquant®, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, chromate, Spectroquant®, Supelco®
Supplier: Merck
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Sodium hypochlorite (14% Cl₂) in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Test kits, photometric, copper, LCK cuvette tests
Supplier: Hach
Powder pillows, ammonia
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach