2,2-BIS(4-HYDROXY-3-METHYLPHENYL)PROPANE 1 * 25 g
Catalog # APOSOR17687-25G
Supplier: Apollo Scientific
2,2-BIS(4-HYDROXY-3-METHYLPHENYL)PROPANE 1 * 25 g
Catalog # APOSOR17687-25G
Supplier: Apollo Scientific
Frequently Bought Together
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
2,3-Dimercapto-1-propanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
Toluene ≥99.7%, CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Water for analysis
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
Supplier: Thermo Fisher Scientific
Acetone ≥99%
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals