HYDRAZINE-1-CARBOHYDRAZONOHYDRAZIDE HYDR 1 * 5 g
Catalog # APOSOR29342-5G
Supplier: Apollo Scientific
HYDRAZINE-1-CARBOHYDRAZONOHYDRAZIDE HYDR 1 * 5 g
Catalog # APOSOR29342-5G
Supplier: Apollo Scientific
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
DIMETHYLSULFOXIDE PURE 1 * 5 L
Supplier: Lach-ner S.R.O.
Acetone reagent ≥99.5% ACS
Supplier: Merck
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
N,N-DIMETHYLFORMAMIDE AR | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND