7,8-DIHYDROXYFLAVONE 1 * 5 g
Catalog # APOSOR52179-5G
Supplier: Apollo Scientific
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chloroform 99.0-99.6% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., VOLUSOL® titration solvent
Supplier: VWR Chemicals
Toluen ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
4-Methoxyphenylhydrazinium chloride ≥99%
Supplier: Thermo Fisher Scientific
(3-Methoxyphenyl)hydrazine hydrochloride 98%
Supplier: Apollo Scientific
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals