2-BROMO-3-BUTOXY-6-FLUOROPHENYLBORONIC A 1 * 250 mg
Catalog # APOSPC412046-250MG
Supplier: Apollo Scientific
2-BROMO-3-BUTOXY-6-FLUOROPHENYLBORONIC A 1 * 250 mg
Catalog # APOSPC412046-250MG
Supplier: Apollo Scientific
Frequently Bought Together
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
Pyridine ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane ≥99% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide ≥99.7% for HPLC
Supplier: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific
Glycerine, Sigma-Aldrich®
Supplier: Merck
Benzyl alcohol ≥99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals