TERT-BUTYL 5-CHLOROSPIRO[INDOLINE-3,4'-P 1 * 250 mg
Catalog # APOSOR315444-250MG
Supplier: Apollo Scientific
TERT-BUTYL 5-CHLOROSPIRO[INDOLINE-3,4'-P 1 * 250 mg
Catalog # APOSOR315444-250MG
Supplier: Apollo Scientific
Frequently Bought Together
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
METHANOL SUITABLE FOR HPLC >99.9% 1 * 2,5 L
Supplier: Merck
1-Propanol ACS for spectroscopy
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem