ETHYL 4-OXOPIPERIDINE-3-CARBOXYLATE HYDR 1 * 100 g
Catalog # APOSOR301044-100G
Supplier: Apollo Scientific
ETHYL 4-OXOPIPERIDINE-3-CARBOXYLATE HYDR 1 * 100 g
Catalog # APOSOR301044-100G
Supplier: Apollo Scientific
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Xylene (mixture of isomers), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Diiodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Bromine 99.6% for analysis
Supplier: Thermo Fisher Scientific
Glycerine, Hi-LR™
Supplier: HIMEDIA
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific