RUTHENIUM(IV) OXIDE HYDRATE, RU 58% 1 * 1 g
Catalog # APOSIN3063-1G
Supplier: Apollo Scientific
Frequently Bought Together
Ammonium heptamolybdate tetrahydrate ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium sulphate, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydrogen carbonate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Potassium hexacyanoferrate(III) ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium salicylate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium hypochlorite 10 - 15% available chlorine in aqueous solution
Supplier: Thermo Fisher Scientific