6,8-DIFLUORO-4-METHYLUMBELLIFERYL ?-D-GL 1 * 10 mg
Catalog # APOSBICL2168-10MG
Supplier: Apollo Scientific
6,8-DIFLUORO-4-METHYLUMBELLIFERYL ?-D-GL 1 * 10 mg
Catalog # APOSBICL2168-10MG
Supplier: Apollo Scientific
Frequently Bought Together
n-Pentane ≥99.5% (GC, on anhydrous basis), PESTINORM® Supra Trace
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 60…80 °C, EMSURE® for analysis, Supelco®
Supplier: Merck
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific