FMOC-(R)-3-AMINO-4-(4-T-BUTYL-PHENYL)-BU 1 * 250 mg
Catalog # APOSOR930345-250MG
Supplier: Apollo Scientific
FMOC-(R)-3-AMINO-4-(4-T-BUTYL-PHENYL)-BU 1 * 250 mg
Catalog # APOSOR930345-250MG
Supplier: Apollo Scientific
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Toluene, anhydrous (max. 0.005% H₂O) ≥99.8%, AnalaR NORMAPUR® for synthesis
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
1-Butanol ≥99%
Supplier: Thermo Fisher Scientific