BARIUM IODIDE DIHYDRATE 1 * 100 g
Catalog # APOSIN1300-100G
Supplier: Apollo Scientific
Frequently Bought Together
Disodium hydrogen phosphate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Barium sulphate ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide 1 mol/l (1 N) in aqueous solution
Supplier: Lach-ner S.R.O.
Sodium hydroxide, pellets Ph. Eur., J.T.Baker®
Supplier: Avantor
Magnesium chloride, anhydrous 99%
Supplier: Thermo Fisher Scientific
Lithium bromide, anhydrous 99+% for analysis
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99%
Supplier: Thermo Fisher Scientific