5-(METHOXYMETHYLENE)-2,2-DIMETHYL-1,3-DI 1 * 25 g
Catalog # APOSOR951532-25G
Supplier: Apollo Scientific
5-(METHOXYMETHYLENE)-2,2-DIMETHYL-1,3-DI 1 * 25 g
Catalog # APOSOR951532-25G
Supplier: Apollo Scientific
Frequently Bought Together
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
2-Propanol ≥99.0% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol eurodenatured ≥80%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Acetone 99.8% for HPLC
Supplier: Thermo Fisher Scientific