2-(2-PYRROLYL)ETHYLAMINE 1 * 1 g
Catalog # APOSOR952802-1G
Supplier: Apollo Scientific
Frequently Bought Together
Acetonitrile ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Petroleum spirit, 60…80 °C
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
4-(Methylamino)phenol sulphate 99%, extra pure
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Water, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Anisole 99%, Reagent Grade
Supplier: Honeywell Chemicals
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific