5-(AMINOMETHYL)-1-ETHYL-3-METHYL-1H-PYRA 1 * 250 mg
Catalog # APOSOR103412-250MG
Supplier: Apollo Scientific
5-(AMINOMETHYL)-1-ETHYL-3-METHYL-1H-PYRA 1 * 250 mg
Catalog # APOSOR103412-250MG
Supplier: Apollo Scientific
Frequently Bought Together
Methanol ≥99.9%, PESTINORM® for purge and trap GC analysis
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Benzyl alcohol, EMSURE® for analysis, Supelco®
Supplier: Merck
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Water, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Supplier: VWR Chemicals
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.7% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals