METHYL 2,3-O-ISOPROPYLIDENE-6-O-TOSYL-?- 1 * 250 mg
Catalog # APOSBICL2442-250MG
Supplier: Apollo Scientific
METHYL 2,3-O-ISOPROPYLIDENE-6-O-TOSYL-?- 1 * 250 mg
Catalog # APOSBICL2442-250MG
Supplier: Apollo Scientific
Frequently Bought Together
Acetonitrile ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
2-Propanol ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Hexane (mixture of isomers) 99+% for HPLC
Supplier: Thermo Fisher Scientific