(4-ETHOXY-2,3-DIFLUOROPHENYL)(METHYL)SUL 1 * 10 g
Catalog # APOSPC99091-10G
Supplier: Apollo Scientific
(4-ETHOXY-2,3-DIFLUOROPHENYL)(METHYL)SUL 1 * 10 g
Catalog # APOSPC99091-10G
Supplier: Apollo Scientific
Frequently Bought Together
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Diethyl ether stabilised, EMPLURA®, Supelco®
Supplier: Merck
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Supplier: Merck
Formamide ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®
Supplier: Merck
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Pyridine, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific