FMOC-ASP(OTBU)-CYS(PSI(ME,ME)PRO)-OH 1 * 250 mg
APOSOR36452-250MG
Frequently Bought Together
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
(±)-2-Butanol ≥99% analytical reagent
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals