TpH 9.000 @ 25°C Millesimal Calibration Buffer, 500 mL 1 * 500 mL
Katalogové číslo HANNHI6009
Dodavatel: Hanna
TpH 9.000 @ 25°C Millesimal Calibration Buffer, 500 mL 1 * 500 mL
Katalogové číslo HANNHI6009
Dodavatel: Hanna
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Acetone ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
Dodavatel: Merck
2-Propanol ≥98%, TECHNICAL
Dodavatel: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Dodavatel: VWR Chemicals