RIVER WATERS TRACE ELEMENTS 1 * 200 mL
Catalog # PROONRC-SLRS-6
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sulphur trioxide pyridine complex for synthesis
Supplier: Merck
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
1-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Klercide™ 70/30 Alcohols, Sterile
Supplier: ECOLAB CONTAMINATION CONTROL
Acetylacetone, EMSURE® for analysis, Supelco®
Supplier: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
1,4-Dioxane, extra dry 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific