ContactUsLinkComponent
Mettler Toledo InPro 3253 Liquid Filled Combination REDOX/pH Electrode with Sterilization Resistant Glass and Automatic Temperature Compensation (PT1000) Rugged IP68 Rated Quick Connect Variopin Connector. 425 mm Length. Includes Cable 2700-0358. 1 * 1 items
Catalog # DISI2800-1230
Supplier:  DISTEK
Mettler Toledo InPro 3253 Liquid Filled Combination REDOX/pH Electrode with Sterilization Resistant Glass and Automatic Temperature Compensation (PT1000) Rugged IP68 Rated Quick Connect Variopin Connector. 425 mm Length. Includes Cable 2700-0358. 1 * 1 items
Catalog # DISI2800-1230
Supplier:  DISTEK
Frequently Bought Together