OLEANDOMYCIN 1000µG/ML IN ACETONITRILE 1 * 1 mL
Catalog # EHERA15725990AL-10
Supplier: EHRENSTORFER
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Density standard, water
Supplier: Mettler - Toledo
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Butane-1,3-diol, Reference Standard
Supplier: USP
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
VWR®, pH-Indicator Paper Congo Red Reag. Ph. Eur. 1022002
Supplier: VWR Chemicals
Tin(II) chloride 30% in hydrochloric acid 20% Reag. Ph. Eur. 1085001
Supplier: VWR Chemicals
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
pH buffers, colour coded, reference standards, Supelco®
Supplier: Merck