AEROMETER WITHOUT THERM. APPROX 180MM cl. H 1.250-1.350 in 0.005 g/cm3, Tp.20C, m.Fa., verifiable 1 * 1 items
Catalog # GERI30818
Supplier: GERING
AEROMETER WITHOUT THERM. APPROX 180MM cl. H 1.250-1.350 in 0.005 g/cm3, Tp.20C, m.Fa., verifiable 1 * 1 items
Catalog # GERI30818
Supplier: GERING
Frequently Bought Together
Short hydrometers
Supplier: Alla France
Milk butyrometer, Gerber's method, VTR glass, s.r.o.
Supplier: Kavalier
Gay-Lussac alcohol meters
Supplier: Alla France
Standard hydrometers, M100
Supplier: Alla France
Burettes, lateral stopcock, class AS, Schellbach
Supplier: witeg Labortechnik
Graduated pipettes for encymatic analysis, Class AS
Supplier: witeg Labortechnik
Volumetric flasks, class A, USP, white markings
Supplier: HIRSCHMANN
Bulb pipettes, one mark, class B
Supplier: HIRSCHMANN
Specific Gravity Hydrometers
Supplier: Amarell
Measuring cylinders, tall form, class A, borosilicate glass
Supplier: GLASSCO
HYDROMETER SOIL ASTM152H-62 -5+60:1g/l 1 * 1 items
Supplier: Amarell
Graduated pipettes, conformity certified, COLOR CODE
Supplier: TECHNOSKLO
Density thermo-hydrometers
Supplier: Alla France
Plastic bases and safety rings for measuring cylinders
Supplier: HIRSCHMANN
Measuring cylinders, tall form, class B, borosilicate glass
Supplier: Kavalier
Volumetric flasks, class A, blue markings
Supplier: HIRSCHMANN
Graduated pipettes, with piston, FORTUNA®
Supplier: Poulten & Graf