ContactUsLinkComponent
MasterSense® MP30 Pressure Sensor with 1¹/₂" Sanitary Tri-Clamp Process Connection
  MFLX07526-82
CE Compliant
IP65 washdown
undefined
MasterSense® MP30 Pressure Sensor with 1¹/₂" Sanitary Tri-Clamp Process Connection
  MFLX07526-82
CE Compliant
IP65 washdown
High accuracy pressure transducers for plug and play connectivity with Masterflex® MasterSense® pump systems.
  • Add critical pressure monitoring to your workflow
  • All-stainless welded body and wetted parts
  • Rugged design for shock protection and excellent long-term stability
  • CE certificated
  • Sanitary models are designed for clean-in-place and sterilize-in-place
  • 0 to 30 psi
  • 4 to 20 mA output
  • CE Compliant
  • IP 65
MasterSense® MP30 pressure transducers utilize a thin-film-on-steel measurement principle to provide an accurate 0 to 30 psi output directly to MasterSense® pump systems. Sensors include a six foot cable with AUX plug adaptor for easy plug and play.
Designed for use with a 1¹/₂" sanitary process tee connection and gasket (not included)
Install downstream of pump to detect pressure changes in your process. Perfect for tangential-Flow filtration, depth filtration, harvest, bulk transfer, chromatography and fill applications. Choose the model with the appropriate process connection for your application.
Sensor MFLX07526-82 can be coupled with MFLX31203-33 single use tee and MFLX31201-95 single-use clamp for a gamma sterilizable flow path.
Sensor MFLX07526-82 can be coupled with MFLX31203-33 single use tee and MFLX31201-95 single-use clamp for a gamma sterilizable flow path. Model MFLX07526-81 ³/₄" sanitary connection not recommended for applications with high temperature variations.
 : CE Certified.
  • Accuracy:
    ±0,5% full scale (0,15 psi) at 25 °C
  • Max. pressure:
    30 psi
  • Temperature range:
    −40...750 °F (media)
  • Description:
    Masterflex® MasterSense® MP30 Pressure Sensor with 1¹/₂" Sanitary Tri-Clamp Process Connection, for MasterSense® Pump Systems
  • Seq:
    03
  • Environmentally Preferable:
  • Wetted materials:
    316 Stainless steel
  • Connection type:
    1¹/₂" sanitary tri-clamp
  • Pk:
    1
  • Included accessories:
    6' cable
Frequently Bought Together