InfinityLab Poroshell 120 Phenyl-Hexyl, 2.1 x 50 mm, 1.9 um, narrow bore LC column, with column ID. Specifications:- Carbon Load: 8 %. Endcapped: Yes. Guard Column: No. Hardware: Conventional. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab 1 * 1 items
Catalog # HEWL699675-912
Supplier: Agilent
Frequently Bought Together