ContactUsLinkComponent
InfinityLab Poroshell 120 HILIC-Z, 2.1 x 150 mm, 2.7 um, PEEK-lined. Specifications:- Endcapped: No. Guard Column: No. Hardware: PEE K Lined SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 8 1 * 1 items
Catalog # HEWL673775-924
Dodavatel:  Agilent
undefined
InfinityLab Poroshell 120 HILIC-Z, 2.1 x 150 mm, 2.7 um, PEEK-lined. Specifications:- Endcapped: No. Guard Column: No. Hardware: PEE K Lined SS. Inner Diameter (ID): 2.1 mm. LC Column Brand: InfinityLab Poroshell 120. LC Column Maximum Temperature: 8 1 * 1 items
Catalog # HEWL673775-924
Dodavatel:  Agilent
Frequently Bought Together