ContactUsLinkComponent
Specifications:- Filter Assembly Material: Stainless Steel. Filter Material: Stainless Steel. Inner Diameter (ID): 4.6 mm. LC Applic ation Compatibility: Analytical. LC Module Type Compatibility: Autosampler Pump Column Compartment. LC Supplies Brand 1 * 5 items
Catalog # HEWL5067-1613
Supplier:  Agilent
Specifications:- Filter Assembly Material: Stainless Steel. Filter Material: Stainless Steel. Inner Diameter (ID): 4.6 mm. LC Applic ation Compatibility: Analytical. LC Module Type Compatibility: Autosampler Pump Column Compartment. LC Supplies Brand 1 * 5 items
Catalog # HEWL5067-1613
Supplier:  Agilent
Frequently Bought Together