ContactUsLinkComponent
Dropping point thermomer belohde, similar DIN, enclosed scale, +100+230:1C, capillary prismatic colourless, gallium filled, 240x10mm, total immersion,Richter-top finish without button, government verified, with Verification Certificate with 3 test p 1 * 1 items
Catalog # AMARM235004-GA-VC
Supplier:  Amarell
Dropping point thermomer belohde, similar DIN, enclosed scale, +100+230:1C, capillary prismatic colourless, gallium filled, 240x10mm, total immersion,Richter-top finish without button, government verified, with Verification Certificate with 3 test p 1 * 1 items
Catalog # AMARM235004-GA-VC
Supplier:  Amarell
Frequently Bought Together