Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]STA-ON TISSUE SECTION ADHESIVE 1 * 3,8 L
Catalog # LEIC3803107
Supplier:
LEICA MICROSYSTEMS
Expand image
[EN]STA-ON TISSUE SECTION ADHESIVE 1 * 3,8 L
Catalog # LEIC3803107
Supplier:
LEICA MICROSYSTEMS
Frequently Bought Together
Ferrous Ammonium Sulphate 0.1M Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
Accessories for photometric test kits, NANOCONTROL
Supplier: MACHEREY-NAGEL
AB 548820 4-[(4-Hydroxyphenyl)diazenyl] benzenesulfonic acid CAS 2918-83-4 1g 1 * 1 g
Supplier: abcr