Reagecon 251.3 mS/cm Conductivity Standard at 25C 1 * 500 mL
Catalog # REGECS2513M
Supplier: REAGECON
Reagecon 251.3 mS/cm Conductivity Standard at 25C 1 * 500 mL
Catalog # REGECS2513M
Supplier: REAGECON
Frequently Bought Together
Cell culture salts, salt solutions and buffers
Supplier: Biowest
VWR®, Nitrile Gloves
Supplier: VWR Collection
Bottle-top dispensers, Dispensette® S Organic
Supplier: Brand
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals