ContactUsLinkComponent
COD Set-Up MD 600 ST-COD sealed tubes ready for use Suppression of chloride interference up to 1000 mg/L (LR & MR) up to 10000 mg/L(HR) Mercury free tube tests,in absence of chloride interference 4 ranges: – Low range: 3 - 150 mg/L, meets ISO 15705:2 1 * 1 items
Catalog # LOVI214040
Supplier:  Lovibond
COD Set-Up MD 600 ST-COD sealed tubes ready for use Suppression of chloride interference up to 1000 mg/L (LR & MR) up to 10000 mg/L(HR) Mercury free tube tests,in absence of chloride interference 4 ranges: – Low range: 3 - 150 mg/L, meets ISO 15705:2 1 * 1 items
Catalog # LOVI214040
Supplier:  Lovibond
Frequently Bought Together