Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Petri dish in polystyrene 65x14,5 mm Contact triple vents easy open double bag sterile 1 * 800 items
391-0680
:
VWR Collection
Petri dish in polystyrene 65x14,5 mm Contact triple vents easy open double bag sterile 1 * 800 items
391-0680
:
VWR Collection
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR® IPASEPT® 70 Cleaning Solution
: VWR Chemicals
LPT DILUTION BROTH BASE 1 * 500 g
: LIOFILCHEM