DEXTROSE 1AQ MULTICOMP CGMP 1 * 12 kg
Catalog # 1915-37
Supplier: Avantor
Frequently Bought Together
Diethylene glycol monomethyl ether, Sigma-Aldrich®
Supplier: Merck
Acetonitrile ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
N-Methyl-2-pyrrolidone (NMP) 99%, Reagent Grade
Supplier: Honeywell Chemicals
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Chloroform ≥99% stabilised ethanol-free
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Ethylene glycol, Sigma-Aldrich®
Supplier: Merck
Ethylene glycol 99.5% for analysis
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Supplier: Thermo Fisher Scientific
Ethanol denatured absolute ≥98% (by GC) (denaturated with 1% MEK and 0.001% Bitrex®)
Supplier: Honeywell Chemicals
2-Propanol USP, J.T.Baker®
Supplier: Avantor