DEXTROSE 1AQ MULTICOMP CGMP 1 * 500 g
Catalog # 1915-31
Supplier: Avantor
Frequently Bought Together
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethylene glycol for synthesis
Supplier: Merck
Diethyl ether stabilised, TECHNICAL, rectified
Supplier: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 80…110 °C, TECHNICAL
Supplier: VWR Chemicals
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥98%, TECHNICAL
Supplier: VWR Chemicals
Dimethyl sulphoxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Triethylamine for HPLC
Supplier: Thermo Fisher Scientific
Iodomethane, Sigma-Aldrich®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals