ContactUsLinkComponent
Tischautoklav / Table top autoclave / HMT-300MB Chamber volume (l): 40 Chamber dimensions O x D (mm): 300 x 570 Outside dimensions W x D x H (mm): 620 x 766 x 489 Working temperature (C): 121 - 135 Power (kW): 3.3 Power connection AC 220 V / 240 V N 1 * 1 items
Catalog # HMCA22021
Supplier:  HMC Europe GmbH
Tischautoklav / Table top autoclave / HMT-300MB Chamber volume (l): 40 Chamber dimensions O x D (mm): 300 x 570 Outside dimensions W x D x H (mm): 620 x 766 x 489 Working temperature (C): 121 - 135 Power (kW): 3.3 Power connection AC 220 V / 240 V N 1 * 1 items
Catalog # HMCA22021
Supplier:  HMC Europe GmbH
Frequently Bought Together