POTASSIUM HYDROXIDE ACS REAGENT >85% 1 * 6 items
Catalog # SIAL221473-6X500G
Supplier: Merck
Frequently Bought Together
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
N-Ethyldiisopropylamine, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Butanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Isobutyronitrile 99%
Supplier: Thermo Fisher Scientific