Lens Tube UV, with lens, Variable bandwidth 1 * 1 items
Catalog # MAPD810703
Supplier: MAPADA
Frequently Bought Together
certificate of analysis for Aqua 90 1 * 1 items
Supplier: LAUDA
Assembled pump NaOH 230V 1 * 1 items
Supplier: VELP SCIENTIFIC
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
Viton gasket standard, 6 pcs. 1 * 6 items
Supplier: BUCHI
Protection GF1/18 1 * 1 items
Supplier: Thermo Scientific
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
FELT RING 1 * 1 items
Supplier: IKA
Screw Cap Gl-10 1 * 1 items
Supplier: BUCHI
Indexing disc (Reax top) 1 * 1 items
Supplier: Heidolph Instruments GmbH & Co.KG
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
Temperature sensor set 1 * 1 items
Supplier: Heidolph Instruments GmbH & Co.KG
MOTOR BRUSH AND SPRING 1 * 1 items
Supplier: BELLCO GLASS
PCB RCT BASIC 230V 1 * 1 items
Supplier: IKA
Single channel pipettes, mechanical, variable volume, PIPETMAN®
Supplier: GILSON
PUSHER LOW 100-1000L F1 LOCKED 1 * 1 items
Supplier: Thermo Fisher Scientific