1-[[9-[2-Hydroxy-3-[[2-(2-methoxy- phenoxy)ethyl]amino]propyl]-9H-carbazol-4- yl]oxy]-3-[[2-(2- 1 * 25 mg
PROOMM0291.01-0025
1-[[9-[2-Hydroxy-3-[[2-(2-methoxy- phenoxy)ethyl]amino]propyl]-9H-carbazol-4- yl]oxy]-3-[[2-(2- 1 * 25 mg
PROOMM0291.01-0025
Frequently Bought Together
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
: VWR Chemicals
pH buffer solutions
: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Diphenylamine 1 g/l in sulphuric acid R Reag. Ph. Eur. 1032101
: VWR Chemicals
Hydrochloric acid 1% Reag. Ph. Eur.
: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
: VWR Chemicals
Water Reag. Ph. Eur. 1095502 carbon dioxide-free
: VWR Chemicals
UV/Visible standards, Supelco®
: Merck
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
: HAMILTON BONADUZ
Benzoic acid, Supelco®
: Merck