ContactUsLinkComponent
CHROMABOND QuEChERS Mix L, Diamino/Carbon clean-up mix, consisting of:, - 900 mg MgSO4, - 300 mg CHROMABOND Diamino, - 150 mg CHROMA BOND Carbon, individually weighed and filled into, 15 mL centrifuge tubes (PP) with screw cap (PE), pack of 50 1 * 135 mg
Catalog # MANA7300008
Supplier:  MACHEREY-NAGEL
CHROMABOND QuEChERS Mix L, Diamino/Carbon clean-up mix, consisting of:, - 900 mg MgSO4, - 300 mg CHROMABOND Diamino, - 150 mg CHROMA BOND Carbon, individually weighed and filled into, 15 mL centrifuge tubes (PP) with screw cap (PE), pack of 50 1 * 135 mg
Catalog # MANA7300008
Supplier:  MACHEREY-NAGEL
Frequently Bought Together