N-Bromosuccinimide 98%, for Synthesis 1 * 100 g
Catalog # 91314.180
Supplier: VWR Chemicals
Frequently Bought Together
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane, extra dry over Molecular Sieve 99.8% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
1,4-Dioxane 99+% ACS
Supplier: Thermo Fisher Scientific
1-Propanol ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Formamide 99.5+%, extra pure
Supplier: Thermo Fisher Scientific
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem
Ethyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific