TETRAFLUOROBORIC ACID, 48 WT. % SOLUTION 1 * 25 g
Catalog # SIAL207934-25G
Supplier: Merck
Frequently Bought Together
Pesticide standard, Azoxystrobin, reference material, Honeywell Fluka™
Supplier: Honeywell Chemicals
Sodium thiosulphate, anhydrous 98.5%, extra pure
Supplier: Thermo Fisher Scientific
Sodium hydrogen carbonate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Potassium chloride ≥99.5% (by argentometric titration), Puriss. p.a.
Supplier: Honeywell Chemicals
Sodium hydrogen carbonate ≥99%
Supplier: Thermo Fisher Scientific
Ammonium acetate ≥97%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
[(2-DI-TERT-BUTYLPHOSPHINO-2',4',6'-TRII 1 * 5 g
Supplier: Apollo Scientific
Lead(II) carbonate basic, extra pure
Supplier: Thermo Fisher Scientific
Sodium tetraphenylborate, Sigma-Aldrich®
Supplier: Merck